Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Oxytriptan (L-5-Hydroxytryptophan)
SDP

Catalog No. 14185610 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-610 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-610 Supplier Medisca Supplier No. 071903
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxytriptan (L-5-Hydroxytryptophan) is a white to off-white hygroscopic powder. Alternative name(s): oxitriptan, l-5HTP

Oxytriptan (L-5-Hydroxytryptophan) is a white to off-white hygroscopic powder.

Alternative name(s): oxitriptan, l-5HTP

Specifications

Chemical Name or Material Oxytriptan (L-5-Hydroxytryptophan)
Packaging Bottle, 30mL, Amber Glass, Wide Mouth Packer, 28-400
CAS 4350-09-8
Type Drug Standard
Molecular Weight (g/mol) 220.2268
Quantity 5 g
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.