Learn More
ALPHATHERA oYo-Link DBCO Antibody Labeling Reagent - Labels 1mg
SDP
Catalog No. NC2147699
Encompass
oYo-Link DBCO enables site-specific labeling to the heavy chain of compatible antibodies with up to two DBCO labels per antibody. oYo-Link enables the click reaction to take place prior to antibody labeling which results in highly efficient click reactions. Simply mix oYo-Link DBCO with desired antibody followed by photocrosslinking for conjugation in 30-sec hands on time, 2 hours total.
Catalog No. NC2147699 Supplier ALPHATHERA Supplier No. AT30031000
oYo-Link DBCO enables site-specific labeling to the heavy chain of compatible antibodies with up to two DBCO labels per antibody. oYo-Link enables the click reaction to take place prior to antibody labeling which results in highly efficient click reactions. Simply mix oYo-Link DBCO with desired antibody followed by photocrosslinking for conjugation in 30-sec hands on time, 2 hours total.