Learn More
ALPHATHERA oYo-Link mIgG1 Azide Antibody Labeling Reagent - Labels 100ug mIgG1 Ab
SDP
Catalog No. NC2112213
Encompass
oYo-Link Azide enables site-specific labeling of azide to the heavy chain of compatible antibodies with up to two Azide labels per antibody. oYo-Link enables the click reaction to take place prior to antibody labeling which results in highly efficient click reactions.Simply mix oYo-Link Azide with your desired antibody followed by photocrosslinking for conjugation in 30-seconds hands on time, 2 hours total. Photocrosslinking can be carried out in most buffers, including those containing amine-containing molecules (e.g. TRIS, glycine) or storage proteins
This item is not returnable. View return policy
Catalog No. NC2112213 Supplier ALPHATHERA Supplier No. AT3002MIGG1100
oYo-Link Azide enables site-specific labeling of azide to the heavy chain of compatible antibodies with up to two Azide labels per antibody. oYo-Link enables the click reaction to take place prior to antibody labeling which results in highly efficient click reactions.Simply mix oYo-Link Azide with your desired antibody followed by photocrosslinking for conjugation in 30-seconds hands on time, 2 hours total. Photocrosslinking can be carried out in most buffers, including those containing amine-containing molecules (e.g. TRIS, glycine) or storage proteins