Learn More
ALPHATHERA oYo-Link Single-Biotin Antibody Labeling Reagent- Labels 500ug
SDP
Catalog No. NC2221693
Encompass
oYo-Link Single-Biotin enables site-specific labeling of compatible antibodies with up to two labels of Biotin for antibody surface immobilization on Streptavidin-coated surfaces. Site-specific immobilization ensures proper antibody orientation, which can improve sensitivity, stability and longevity of capture antibodies for ELISA. Simply mix the oYo-Link Single-Biotin with desired antibody followed by photocrosslinking for conjugation in 30-sec hands on time, 2 hours total.
This item is not returnable. View return policy
Catalog No. NC2221693 Supplier ALPHATHERA Supplier No. AT4001500
oYo-Link Single-Biotin enables site-specific labeling of compatible antibodies with up to two labels of Biotin for antibody surface immobilization on Streptavidin-coated surfaces. Site-specific immobilization ensures proper antibody orientation, which can improve sensitivity, stability and longevity of capture antibodies for ELISA. Simply mix the oYo-Link Single-Biotin with desired antibody followed by photocrosslinking for conjugation in 30-sec hands on time, 2 hours total.