Learn More
Electron Microscopy Sciences Veco Oyster Hexagonal DH75/300, 75/300, Copper
SDP
Catalog No. 5027600
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 333/ µm
Hole: 293/ µm
Bar: 50/ µm
Catalog No. 50-276-00 Supplier Electron Microscopy Sciences Supplier No. D753HCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 333/ µm
Hole: 293/ µm
Bar: 50/ µm