Learn More
Electron Microscopy Sciences Veco Oyster Hexagonal DH75, 75 mesh Copper Grid
SDP
Catalog No. 5027592
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 333 µm
Hole: 283 µm
Bar: 50 µm
Catalog No. 50-275-92 Supplier Electron Microscopy Sciences Supplier No. D75HCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 333 µm
Hole: 283 µm
Bar: 50 µm