Learn More
Electron Microscopy Sciences Veco Oyster Slot Grid 2000 x 1000 µm, Nickel
SDP
Catalog No. 5027602
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Catalog No. 50-276-02 Supplier Electron Microscopy Sciences Supplier No. DL2010NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel