Learn More
Enzo Life Sciences Ozagrel (10mg). CAS: 82571-53-7
SDP
Supplier:  Enzo Life Sciences BMLEI3780010
Encompass_Preferred
Potent and selective thromboxane A2 synthase inhibitor (IC50=11nM). Purity: ≥98%. Long Term Storage: .
Catalog No. 50-201-0760