Learn More
Medchemexpress LLC HY-B0428 10mg Medchemexpress, Ozagrel CAS:82571-53-7 Purity:>98%
SDP
Catalog No. 502082530
Encompass_Preferred
Medchemexpress, HY-B0428 10mg Ozagrel CAS:82571-53-7 Ozagrel (OKY-046) is a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2530 Supplier Medchemexpress LLC Supplier No. HYB042810MG
Medchemexpress, HY-B0428 10mg Ozagrel CAS:82571-53-7 Ozagrel (OKY-046) is a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.