Learn More
Apexbio Technology LLC Ozagrel 82571-53-7 10mg
SDP
Catalog No. 502240734
Encompass_Preferred
Ozagrel (CAS 82571-53-7) is a selective inhibitor of thromboxane A synthase an enzyme involved in the conversion of prostaglandin H to thromboxane A a potent mediator of platelet aggregation and vasoconstriction Ozagrel demonstrates high inhibitory potency with an IC of 4 nM indicating strong activity at low concentrations This compound is widely utilized in research to investigate thromboxane-dependent signaling pathways vascular biology and platelet function as well as in studies exploring potential therapeutic interventions targeting thrombotic and cardiovascular disorders
This item is not returnable. View return policy
Catalog No. 50-224-0734 Supplier Apexbio Technology LLC Supplier No. A434410
Ozagrel (CAS 82571-53-7) is a selective inhibitor of thromboxane A synthase an enzyme involved in the conversion of prostaglandin H to thromboxane A a potent mediator of platelet aggregation and vasoconstriction Ozagrel demonstrates high inhibitory potency with an IC of 4 nM indicating strong activity at low concentrations This compound is widely utilized in research to investigate thromboxane-dependent signaling pathways vascular biology and platelet function as well as in studies exploring potential therapeutic interventions targeting thrombotic and cardiovascular disorders