Learn More
AdipoGen Ozagrel
SDP
Catalog No. 502560298
Encompass_Preferred
Chemical. CAS 82571-53-7. Formula C13H12N2O2. MW 228.2. Synthetic. Ozagrel, an antithrombic drug, is a potent and selective TXA Synthase thromboxane A2 TXAS inhibitor. Acts as a selective inhibitor of TXAS with an IC50=11nM. The beneficial effects of TXAS inhibition by ozagrel include improved motor coordination after experimental stroke, and antihypertensive effects in spontaneously hypertensive rats.
Catalog No. 50-256-0298 Supplier AdipoGen Supplier No. CDXO0141M050
Chemical. CAS 82571-53-7. Formula C13H12N2O2. MW 228.2. Synthetic. Ozagrel, an antithrombic drug, is a potent and selective TXA Synthase thromboxane A2 TXAS inhibitor. Acts as a selective inhibitor of TXAS with an IC50=11nM. The beneficial effects of TXAS inhibition by ozagrel include improved motor coordination after experimental stroke, and antihypertensive effects in spontaneously hypertensive rats.