Learn More
Apexbio Technology LLC Ozagrel HCl 78712-43-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000091066
Encompass_Preferred
Ozagrel hydrochloride (CAS 78712-43-3) is a small molecule inhibitor of thromboxane A2 (TXA2) synthase modulating platelet activity by suppressing TXA2 production In experimental models Ozagrel hydrochloride selectively inhibits TXA2-mediated human platelet aggregation exhibiting an IC50 of approximately 53 12 M This compound is employed in platelet-related assays to investigate the role of TXA2 signaling in coagulation thrombosis and cardiovascular pathophysiology
Catalog No. 50-000-91066 Supplier Apexbio Technology LLC Supplier No. B21165.1
Ozagrel hydrochloride (CAS 78712-43-3) is a small molecule inhibitor of thromboxane A2 (TXA2) synthase modulating platelet activity by suppressing TXA2 production In experimental models Ozagrel hydrochloride selectively inhibits TXA2-mediated human platelet aggregation exhibiting an IC50 of approximately 53 12 M This compound is employed in platelet-related assays to investigate the role of TXA2 signaling in coagulation thrombosis and cardiovascular pathophysiology