Learn More
Medchemexpress LLC Ozagrel hydrochloride | 78712-43-3 | 99.9% | 50 MG
SDP
Catalog No. 5000409335
Encompass_Preferred
Ozagrel hydrochloride (OKY-046 hydrochloride) is a highly selective and orally active thromboxane A2 (TXA2) synthase inhibitor with an IC50 of 11 nM. It exerts anti-platelet aggregation, vasodilation, and anti-inflammatory effects by inhibiting TXA2 production and increasing prostacyclin (PGI2) production. This product is intended for research use only and can be utilized for studies related to ischemic stroke, asthma, and thromboembolic diseases.
  • Highly selective and orally active thromboxane A2 (TXA2) synthase inhibitor
  • Exerts anti-platelet aggregation effects
  • Promotes vasodilation
  • Provides anti-inflammatory effects
  • Inhibits TXA2 production
  • Increases prostacyclin (PGI2) production
  • Useful for research in ischemic stroke
  • Useful for research in asthma
  • Useful for research in thromboembolic diseases
Catalog No. 50-004-09335 Supplier Medchemexpress LLC Supplier No. HYB0428B50MG
Ozagrel hydrochloride (OKY-046 hydrochloride) is a highly selective and orally active thromboxane A2 (TXA2) synthase inhibitor with an IC50 of 11 nM. It exerts anti-platelet aggregation, vasodilation, and anti-inflammatory effects by inhibiting TXA2 production and increasing prostacyclin (PGI2) production. This product is intended for research use only and can be utilized for studies related to ischemic stroke, asthma, and thromboembolic diseases.
  • Highly selective and orally active thromboxane A2 (TXA2) synthase inhibitor
  • Exerts anti-platelet aggregation effects
  • Promotes vasodilation
  • Provides anti-inflammatory effects
  • Inhibits TXA2 production
  • Increases prostacyclin (PGI2) production
  • Useful for research in ischemic stroke
  • Useful for research in asthma
  • Useful for research in thromboembolic diseases