Learn More
Medchemexpress LLC Ozagrel methyl ester | 866157-50-8 | 242.27 | 250 MG
SDP
Catalog No. 5000450070
Encompass_Preferred
Ozagrel methyl ester is a chemical compound primarily used as a drug impurity for research purposes. It is supplied as a solid.
  • For research use only
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year
Catalog No. 50-004-50070 Supplier Medchemexpress LLC Supplier No. 250MGHYW004645
Add to Cart
Add to Cart
Ozagrel methyl ester is a chemical compound primarily used as a drug impurity for research purposes. It is supplied as a solid.
  • For research use only
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year