Learn More
Medchemexpress LLC HY-B0428A 10mg Medchemexpress, Ozagrel (sodium) CAS:189224-26-8 Purity:>98%
SDP
Catalog No. 502082531
Encompass_Preferred
Medchemexpress, HY-B0428A 10mg Ozagrel (sodium) CAS:189224-26-8 Ozagrel sodium (OKY-046 sodium) is a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel sodium is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2531 Supplier Medchemexpress LLC Supplier No. HYB0428A10MG
Medchemexpress, HY-B0428A 10mg Ozagrel (sodium) CAS:189224-26-8 Ozagrel sodium (OKY-046 sodium) is a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel sodium is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.