Learn More
Medchemexpress LLC Ozagrel (sodium) | 189224-26-8 | 99.8% | 1 ML
SDP
Catalog No. 5000409167
Encompass_Preferred
Ozagrel sodium is a highly selective and orally active thromboxane A2 (TXA2) synthase inhibitor with an IC50 of 11 nM. It exerts anti-platelet aggregation, vasodilation, and anti-inflammatory effects by inhibiting the production of TXA2 and increasing the production of prostacyclin (PGI2).
  • Highly selective and orally active
  • Inhibits thromboxane A2 (TXA2) synthase
  • Exerts anti-platelet aggregation effects
  • Promotes vasodilation
  • Possesses anti-inflammatory properties
  • Increases prostacyclin (PGI2) production
  • Suitable for research on ischemic stroke, asthma, and thromboembolic diseases
Catalog No. 50-004-09167 Supplier Medchemexpress LLC Supplier No. HYB0428A10MM/1ML
Ozagrel sodium is a highly selective and orally active thromboxane A2 (TXA2) synthase inhibitor with an IC50 of 11 nM. It exerts anti-platelet aggregation, vasodilation, and anti-inflammatory effects by inhibiting the production of TXA2 and increasing the production of prostacyclin (PGI2).
  • Highly selective and orally active
  • Inhibits thromboxane A2 (TXA2) synthase
  • Exerts anti-platelet aggregation effects
  • Promotes vasodilation
  • Possesses anti-inflammatory properties
  • Increases prostacyclin (PGI2) production
  • Suitable for research on ischemic stroke, asthma, and thromboembolic diseases