Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ozanimod (hydrochloride) | 1618636-37-5 | 98.1% | 1 ML
SDP

Catalog No. 5000232654
Encompass_Preferred

Ozanimod hydrochloride 10 mM in DMSO (1 mL) is a ready-to-use research reagent for studying sphingosine 1-phosphate (S1P) receptor modulation in preclinical and biochemical assays. Supplied as a solution to improve handling and reproducibility. For research use only; not for human use.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot for small-scale experiments.
  • Listed purity 98.1% for consistent experimental results.
  • Molecular weight 440.92 g/mol and formula C23H25ClN4O3.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-002-32654 Supplier Medchemexpress LLC Supplier No. HY12288A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ozanimod hydrochloride 10 mM in DMSO (1 mL) is a ready-to-use research reagent for studying sphingosine 1-phosphate (S1P) receptor modulation in preclinical and biochemical assays. Supplied as a solution to improve handling and reproducibility. For research use only; not for human use.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot for small-scale experiments.
  • Listed purity 98.1% for consistent experimental results.
  • Molecular weight 440.92 g/mol and formula C23H25ClN4O3.
  • Intended for research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.