Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ozanimod hydrochloride | 1618636-37-5 | 98.1% | 10 MG
SDP

Catalog No. 5000230609
Encompass_Preferred

Ozanimod hydrochloride is the hydrochloride salt of ozanimod (RPC-1063), a selective sphingosine 1-phosphate (S1P) receptor modulator supplied for laboratory research use only. It is used in studies of immune modulation and relapsing multiple sclerosis.

  • Purity 98.1%.
  • Molecular weight 440.92.
  • Molecular formula C23H25ClN4O3.
  • White to off-white solid; also available as 10 mM solution in DMSO.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for laboratory research on S1P receptor biology and multiple sclerosis models.

Catalog No. 50-002-30609 Supplier Medchemexpress LLC Supplier No. HY12288A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ozanimod hydrochloride is the hydrochloride salt of ozanimod (RPC-1063), a selective sphingosine 1-phosphate (S1P) receptor modulator supplied for laboratory research use only. It is used in studies of immune modulation and relapsing multiple sclerosis.

  • Purity 98.1%.
  • Molecular weight 440.92.
  • Molecular formula C23H25ClN4O3.
  • White to off-white solid; also available as 10 mM solution in DMSO.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for laboratory research on S1P receptor biology and multiple sclerosis models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.