Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ozanimod hydrochloride | 1618636-37-5 | 98.1% | 100 MG
SDP

Catalog No. 5000230549
Encompass_Preferred

Ozanimod hydrochloride is the hydrochloride salt of ozanimod (RPC-1063), a selective sphingosine 1-phosphate (S1P) receptor modulator used in pharmacological and preclinical research. Supplied as a solid powder and as DMSO solution formats; intended for research use only.

  • Hydrochloride salt form suitable for research applications.
  • CAS number 1618636-37-5.
  • Molecular formula C23H25ClN4O3.
  • Molecular weight 440.92 g/mol.
  • Purity approximately 98.1% by HPLC.
  • Available as solid and as DMSO solution for biochemical assays.
  • For research use only; not for human use.

Catalog No. 50-002-30549 Supplier Medchemexpress LLC Supplier No. HY12288A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ozanimod hydrochloride is the hydrochloride salt of ozanimod (RPC-1063), a selective sphingosine 1-phosphate (S1P) receptor modulator used in pharmacological and preclinical research. Supplied as a solid powder and as DMSO solution formats; intended for research use only.

  • Hydrochloride salt form suitable for research applications.
  • CAS number 1618636-37-5.
  • Molecular formula C23H25ClN4O3.
  • Molecular weight 440.92 g/mol.
  • Purity approximately 98.1% by HPLC.
  • Available as solid and as DMSO solution for biochemical assays.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.