Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ozanimod hydrochloride | 1618636-37-5 | 98.1% | 5 MG
SDP

Catalog No. 5000231025
Encompass_Preferred

Ozanimod hydrochloride is the hydrochloride salt of ozanimod, a selective sphingosine 1-phosphate (S1P) receptor modulator used in research on immune-mediated disorders such as relapsing multiple sclerosis. It is supplied as a white to off-white solid with high purity for in vitro and in vivo pharmacology studies.

  • High purity (98.1%).
  • Molecular weight 440.92 g/mol; formula C23H25ClN4O3.
  • Soluble in DMSO at ≥ 200 mg/mL for stock solutions.
  • Suitable for in vitro and in vivo pharmacology applications.
  • Supplied as a 5 MG quantity.
  • Store sealed at 4°C; in solvent store -80°C for up to 6 months or -20°C for 1 month.

Catalog No. 50-002-31025 Supplier Medchemexpress LLC Supplier No. HY12288A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ozanimod hydrochloride is the hydrochloride salt of ozanimod, a selective sphingosine 1-phosphate (S1P) receptor modulator used in research on immune-mediated disorders such as relapsing multiple sclerosis. It is supplied as a white to off-white solid with high purity for in vitro and in vivo pharmacology studies.

  • High purity (98.1%).
  • Molecular weight 440.92 g/mol; formula C23H25ClN4O3.
  • Soluble in DMSO at ≥ 200 mg/mL for stock solutions.
  • Suitable for in vitro and in vivo pharmacology applications.
  • Supplied as a 5 MG quantity.
  • Store sealed at 4°C; in solvent store -80°C for up to 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.