Learn More
Apexbio Technology LLC Ozanimod (RPC1063), 1306760-87-1, MFCD28386168, 5 mg
SDP
Supplier:  Apexbio Technology LLC B60385
Encompass_Preferred
MF: C23H24N4O3, MW: 404.46, Solubility: 40.4mg/mL in DMSO. Ozanimod (RPC1063) is a high orallybioavailable agonist of the sphingosine-1-phosphate receptor subtypes 1 (S1P1) and 5 (S1P5). S1P receptors belong to the family of G-protein-coupled receptor which contains five subtypes, denoting S1P1-5. S1P receptors are involved in coronary artery caliber,endothelial integrity, cardiac function, and lymphocyte recirculation.
Catalog No. 50-115-2561