Learn More
Medchemexpress LLC Ozenoxacin-d3 (hydrochloride) | 1797984-75-6 | ≥99.0% | 402.89 | C21H19D3ClN3O3 | 10 MG
SDP
Catalog No. 5000286814
Encompass_Preferred
Ozenoxacin-d3 hydrochloride is the deuterium-labeled isotopic standard of ozenoxacin hydrochloride intended for use as an analytical/internal standard in quantitative assays such as LC-MS/MS. Supplied as the hydrochloride salt, it is suitable for method development, validation, and accuracy assessment in trace-level quantitation.
Catalog No. 50-002-86814 Supplier Medchemexpress LLC Supplier No. HY14957AS10MG
Ozenoxacin-d3 hydrochloride is the deuterium-labeled isotopic standard of ozenoxacin hydrochloride intended for use as an analytical/internal standard in quantitative assays such as LC-MS/MS. Supplied as the hydrochloride salt, it is suitable for method development, validation, and accuracy assessment in trace-level quantitation.