Learn More
Medchemexpress LLC Ozenoxacin-d3 (hydrochloride) | 245765-41-7 | ≥99.0% | 402.89 | 5 MG
SDP
Catalog No. 5000360273
Encompass_Preferred
Ozenoxacin-d3 hydrochloride is a deuterium-labeled form of Ozenoxacin hydrochloride. Ozenoxacin hydrochloride is a nonfluorinated quinolone antibacterial that exhibits potent activities against the main microorganisms isolated from skin and soft tissue infections. This compound can be utilized as a tracer or as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits potent activities against microorganisms isolated from skin and soft tissue infections
Catalog No. 50-003-60273 Supplier Medchemexpress LLC Supplier No. HY14957AS5MG
Ozenoxacin-d3 hydrochloride is a deuterium-labeled form of Ozenoxacin hydrochloride. Ozenoxacin hydrochloride is a nonfluorinated quinolone antibacterial that exhibits potent activities against the main microorganisms isolated from skin and soft tissue infections. This compound can be utilized as a tracer or as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits potent activities against microorganisms isolated from skin and soft tissue infections