Learn More
Hach Company Ozone AccuVac Ampules, MR, pk/25
Supplier:  Hach Company 2517025
Encompass
For determination of Ozone by the Indigo method, using AccuVac Ampules. Hach method 8311. Range: 0.01 - 0.75 mg/L O3. Pack of 25 ampules.
Catalog No. 50-199-6169