Learn More
Hach Company Ozone AccuVac Ampules, HR, pk/25
Supplier:  Hach Company 2518025
Encompass
For determination of Ozone by the Indigo method, using AccuVac Ampules. Hach method 8311. Range: 0.01 - 1.50 mg/L O₃. Pack of 25 ampules.
Catalog No. 50-199-6172