Learn More
CHEMetrics™ by AquaPhoenix Ozone/Chlorine Dioxide Refill Kit

Description
- Sample is treated with an excess of potassium iodide.
- Ozone oxidizes the iodide to iodine, and the iodine then oxidizes DPD (N,N-diethyl-p-phenylenediamine) to form a pink colored species in direct proportion to the ozone concentration. Results are expressed as ppm (mg/L) O3. The DPD chemistry can be used to determine the concentration of Total Residual Oxidizers (TRO) in seawater.
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-7404 and K-2705 |
| No. of Tests | 30 |
| Quantity | 30/Box |
| Type | Ozone/Chlorine Dioxide Refill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.