Learn More
Hach Company Hach Recharge Kit - 4 Pre-Filled Cartridges with Premounted Membranes for Orbisphere C1100 Ozone Sensors (33051-xT cap)
Catalog No. NC2123840 Shop All Hach Company Products
Encompass
Hach rechargeable kit containing four pre-filled sensor cartridges with premounted membranes, designed to service Orbisphere C1100 ozone sensors that use the 33051-xT cap. The kit provides the consumables and maintenance accessories needed to replace sensor cartridges and help maintain measurement accuracy.
  • Contains four pre-filled sensor cartridges with premounted membranes.
  • Includes replacement o-rings and cleaning tools for cartridge maintenance.
  • Restores sensor assembly consumables to maintain measurement performance.
  • Supplies multiple replacement cartridges to reduce downtime.
  • Requires matching cap type; verify cap compatibility prior to installation.
This item is not returnable. View return policy
Catalog No. NC2123840 Supplier Hach Company Supplier No. 2956ACT
Hach rechargeable kit containing four pre-filled sensor cartridges with premounted membranes, designed to service Orbisphere C1100 ozone sensors that use the 33051-xT cap. The kit provides the consumables and maintenance accessories needed to replace sensor cartridges and help maintain measurement accuracy.
  • Contains four pre-filled sensor cartridges with premounted membranes.
  • Includes replacement o-rings and cleaning tools for cartridge maintenance.
  • Restores sensor assembly consumables to maintain measurement performance.
  • Supplies multiple replacement cartridges to reduce downtime.
  • Requires matching cap type; verify cap compatibility prior to installation.