Learn More
CTL Scientific Supply Corp OZONE TEST STICKS,PK/24 STRIPS
SDP
Catalog No. 501173301 Shop All CTL Scientific Supply Products
Encompass_Preferred
OZONE TEST STICKS Test strip for the semi-quantitative of ozone in air. 2 packs of 12 test strips. Quick, convenient and reliable onsite ozone testing. Gradation of scale: <90 • 90-150 • 150-210 •>210 µg/m3 O3
Catalog No. 50-117-3301 Supplier CTL Scientific Supply Corp Supplier No. 907362
OZONE TEST STICKS Test strip for the semi-quantitative of ozone in air. 2 packs of 12 test strips. Quick, convenient and reliable onsite ozone testing. Gradation of scale: <90 • 90-150 • 150-210 •>210 µg/m3 O3