Learn More
CHEMetrics™ by AquaPhoenix Ozone Water Quality Visual Test Kit
SDP
Catalog No. 13299634
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-634 30
1 options
Catalog No. 13-299-634 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K7404
Add to Cart
Add to Cart
Ozone visual test kit supplied in a plastic case. Contains everything needed to perform 30 tests.
  • The K-7404 ozone analysis kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low and High Range Comparators, Activator Solution, 25 mL sample cup, and instructions.
Specifications
Analytes Ozone
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 0.60 and 0.6 to 3.0 ppm
Type Water Quality Visual Test Kit