Learn More
Sutter Instruments P-1000 Micropipette Puller
SDP
Catalog No. NC1824200
Encompass_Preferred
New features for the P-1000 include: diagnostic testing of all puller components, built-in error detection of air pressure loss or filament burnout, easy access to ramp test, measurement of jaw temperature, and access to previous pull results with the heat on times for each cycle of the program. In addition, help topics are pre-loaded to assist with on-site troubleshooting and the built-in glossary includes text, pictures, and diagrams explaining the terms used in micropipette fabrication. A rotary dial is offered as an alternative to the keypad for numerical entry.
Catalog No. NC1824200 Supplier Sutter Instruments Supplier No. FGP1000
Add to Cart
Add to Cart
Request a Quote
New features for the P-1000 include: diagnostic testing of all puller components, built-in error detection of air pressure loss or filament burnout, easy access to ramp test, measurement of jaw temperature, and access to previous pull results with the heat on times for each cycle of the program. In addition, help topics are pre-loaded to assist with on-site troubleshooting and the built-in glossary includes text, pictures, and diagrams explaining the terms used in micropipette fabrication. A rotary dial is offered as an alternative to the keypad for numerical entry.