Learn More
Medchemexpress LLC HY-108573 5mg Medchemexpress, P-1075 CAS:60559-98-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085735MG
Encompass_Preferred
Medchemexpress, HY-108573 5mg P-1075 CAS:60559-98-0 P-1075 is a potent activator of sulfonylurea receptor 2-associated ATP-sensitive potassium channels (SUR2-KIR6), with an EC50 value of 45 nM for SUR2B-KIR6 channel activation. P-1075 also P1075 opens mitochondrial K(ATP) channels and generates reactive oxygen species resulting in cardioprotection of rabbit hearts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8577