Learn More
TARGETMOL CHEMICALS INC P 218 1MG
SDP
Catalog No. 502878194
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. P 218 is a novel DHFR inhibitor with antimalarial activity and antifolate effects and can be used to study Buruli ulcer. Purity 99.5%
Catalog No. 50-287-8194 Supplier TARGETMOL CHEMICALS INC Supplier No. T73328L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. P 218 is a novel DHFR inhibitor with antimalarial activity and antifolate effects and can be used to study Buruli ulcer. Purity 99.5%