Learn More
TARGETMOL CHEMICALS INC P 22077 25MG
SDP
Catalog No. 502580820
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. P 22077 (P22077) is an inhibitor of ubiquitin-specific protease USP7 with EC50 of 8.6 uM. It also inhibits the closely related USP47. purity: 98%
Catalog No. 50-258-0820 Supplier TARGETMOL CHEMICALS INC Supplier No. T242425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. P 22077 (P22077) is an inhibitor of ubiquitin-specific protease USP7 with EC50 of 8.6 uM. It also inhibits the closely related USP47. purity: 98%