Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-2281 | 1112994-35-0 | 209.63 | C9H8ClN3O | 10 MG
SDP

Catalog No. 5000228419
Encompass_Preferred

P-2281 is a small-molecule mTOR inhibitor provided for laboratory research use. It shows reported anticancer and anti-inflammatory activity in preclinical studies, has a molecular weight of 209.63 g/mol, and is highly soluble in DMSO. Store protected from light; in solution, long-term storage at -80°C is recommended.

  • Small-molecule inhibitor of mTOR suitable for research applications.
  • Soluble in DMSO at high concentration (125 mg/mL; may require ultrasonic assistance).
  • Molecular weight 209.63 g/mol for dosing and calculation purposes.
  • Stable under recommended storage: short-term at -20°C in solution and long-term at -80°C; protect from light.
  • Intended for in vitro and preclinical research use only.

Catalog No. 50-002-28419 Supplier Medchemexpress LLC Supplier No. HY11870410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P-2281 is a small-molecule mTOR inhibitor provided for laboratory research use. It shows reported anticancer and anti-inflammatory activity in preclinical studies, has a molecular weight of 209.63 g/mol, and is highly soluble in DMSO. Store protected from light; in solution, long-term storage at -80°C is recommended.

  • Small-molecule inhibitor of mTOR suitable for research applications.
  • Soluble in DMSO at high concentration (125 mg/mL; may require ultrasonic assistance).
  • Molecular weight 209.63 g/mol for dosing and calculation purposes.
  • Stable under recommended storage: short-term at -20°C in solution and long-term at -80°C; protect from light.
  • Intended for in vitro and preclinical research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.