Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-2281 | 1112994-35-0 | >98.0% | 209.63 | C9H8ClN3O | 50 MG
SDP

Catalog No. 5000228417
Encompass_Preferred

P-2281 is a small-molecule mTOR inhibitor reported to have anticancer and anti-inflammatory activity. It is supplied as a solid and commonly used in cellular assays to probe mTOR signaling.

  • mTOR inhibitor with demonstrated anticancer and anti-inflammatory effects.
  • CAS 1112994-35-0.
  • Molecular weight 209.63 g/mol.
  • Chemical formula C9H8ClN3O.
  • Purity greater than 98% (as reported).
  • Solid appearance; soluble in DMSO.

Catalog No. 50-002-28417 Supplier Medchemexpress LLC Supplier No. HY11870450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P-2281 is a small-molecule mTOR inhibitor reported to have anticancer and anti-inflammatory activity. It is supplied as a solid and commonly used in cellular assays to probe mTOR signaling.

  • mTOR inhibitor with demonstrated anticancer and anti-inflammatory effects.
  • CAS 1112994-35-0.
  • Molecular weight 209.63 g/mol.
  • Chemical formula C9H8ClN3O.
  • Purity greater than 98% (as reported).
  • Solid appearance; soluble in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.