Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-2281 | 1112994-35-0 | MFCD24392542 | >98.0% | 209.63 g/mol | C9H8ClN3O | 5 MG
SDP

Catalog No. 5000229117
Encompass_Preferred

P-2281 is a synthetic small-molecule research compound reported as an mTOR inhibitor for in vitro studies. It is supplied as a solid, soluble in DMSO, and intended for research use only (not for human or clinical use).

  • CAS number 1112994-35-0
  • Chemical formula C9H8ClN3O
  • Molecular weight 209.63 g/mol
  • Purity >98%
  • Soluble in DMSO (reported ≈125 mg/mL)
  • Typical storage -20 °C to preserve stability
  • Available in small laboratory pack sizes (for example, 1 mg, 5 mg, 10 mg)

Catalog No. 50-002-29117 Supplier Medchemexpress LLC Supplier No. HY1187045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

P-2281 is a synthetic small-molecule research compound reported as an mTOR inhibitor for in vitro studies. It is supplied as a solid, soluble in DMSO, and intended for research use only (not for human or clinical use).

  • CAS number 1112994-35-0
  • Chemical formula C9H8ClN3O
  • Molecular weight 209.63 g/mol
  • Purity >98%
  • Soluble in DMSO (reported ≈125 mg/mL)
  • Typical storage -20 °C to preserve stability
  • Available in small laboratory pack sizes (for example, 1 mg, 5 mg, 10 mg)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.