Learn More
Cole-Parmer Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread
Catalog No. NC1988383 Shop All Cole-Parmer Products
Encompass
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
Catalog No. NC1988383 Supplier Cole-Parmer Supplier No. 0201840
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.