Learn More
Epigentek Inc Epigenase m6A Methylase Activity/Inhibition Assay Kit (Colorimetric) (48 Assays)
SDP
Catalog No. NC1863309
Encompass_Preferred
For measurement of total activity/inhibition of m6A methylases from nuclear extracts or purified enzymes
Catalog No. NC1863309 Supplier Epigentek Inc Supplier No. P901948
For measurement of total activity/inhibition of m6A methylases from nuclear extracts or purified enzymes