Learn More
Electron Microscopy Sciences P.A.W.S. Personal Antimicrobial Wipes
SDP
Catalog No. 50930399
Encompass_Preferred
P.A.W.S. provides sanitizing and extended antimicrobial action on the skin with an individually packaged. Pre-moistened 5"x8" towelette. Great for hands, P.A.W.S. is formulated with PCMX (Chloroxylenol -an antimocrobial agent) and Alcohol and provides the most rapid and greatest reduction of microbial counts on the skin (helping you comply with OSHA Bloodborne Pathogen Regulations). In addition, P.A.W.S. contains Aloe Vera to condition the skin and maintain dermal integrity.
This item is not returnable. View return policy
Catalog No. 50-930-399 Supplier Electron Microscopy Sciences Supplier No. 7057601
P.A.W.S. provides sanitizing and extended antimicrobial action on the skin with an individually packaged. Pre-moistened 5"x8" towelette. Great for hands, P.A.W.S. is formulated with PCMX (Chloroxylenol -an antimocrobial agent) and Alcohol and provides the most rapid and greatest reduction of microbial counts on the skin (helping you comply with OSHA Bloodborne Pathogen Regulations). In addition, P.A.W.S. contains Aloe Vera to condition the skin and maintain dermal integrity.