Learn More
FUJIFILM Biosciences p-Amidinophenylmethanesulf
Catalog No. 5000450228
Encompass_Preferred
p-APMSF is used as a serine protease activity inhibitor Since it specifically suppresses the activity of blood coagulation factor Xa it is used for the study of blood coagulation mechanism Compared to DFP diisopropylfluorophosphate it is easy to handle because it is powder and soluble to water In addition it is reported that serine protease inhibitory activity is several hundred times higher than DFP and PMSF It inhibits the activity of trypsin thrombin plasmin kallikrein and others It does not affect chymotrypsin or acetylcholinesterase For research use only RUO
Catalog No. 50-004-50228 Supplier FUJIFILM Biosciences Supplier No. 01526333
p-APMSF is used as a serine protease activity inhibitor Since it specifically suppresses the activity of blood coagulation factor Xa it is used for the study of blood coagulation mechanism Compared to DFP diisopropylfluorophosphate it is easy to handle because it is powder and soluble to water In addition it is reported that serine protease inhibitory activity is several hundred times higher than DFP and PMSF It inhibits the activity of trypsin thrombin plasmin kallikrein and others It does not affect chymotrypsin or acetylcholinesterase For research use only RUO