Learn More
Medchemexpress LLC P-amidinophenylmethylsulfonylfluoride hydrochloride | 74938-88-8 | MFCD00078902 | 95.0% | 252.69 | C8H10ClFN2O2S | 1 MG
SDP
Catalog No. 5000304071
Encompass_Preferred
p-APMSF hydrochloride is the hydrochloride salt of p-amidinophenylmethylsulfonylfluoride, an irreversible serine protease inhibitor selective for substrates with positively charged side chains such as lysine and arginine. It is provided as a high-purity research reagent for enzymology and biochemical assays.
  • Irreversible serine protease inhibitor selective for positively charged side chains.
  • Provided as the hydrochloride salt to aid solubility in aqueous assays.
  • High purity (95.0%) suitable for research and assay development.
  • Molecular weight 252.69; formula C8H10ClFN2O2S.
  • Offered in small-scale packaging (1 mg) for screening and mechanistic studies.
Catalog No. 50-003-04071 Supplier Medchemexpress LLC Supplier No. HYW0141341MG
p-APMSF hydrochloride is the hydrochloride salt of p-amidinophenylmethylsulfonylfluoride, an irreversible serine protease inhibitor selective for substrates with positively charged side chains such as lysine and arginine. It is provided as a high-purity research reagent for enzymology and biochemical assays.
  • Irreversible serine protease inhibitor selective for positively charged side chains.
  • Provided as the hydrochloride salt to aid solubility in aqueous assays.
  • High purity (95.0%) suitable for research and assay development.
  • Molecular weight 252.69; formula C8H10ClFN2O2S.
  • Offered in small-scale packaging (1 mg) for screening and mechanistic studies.