Learn More
AG Scientific Inc p-Amino Benzoic Acid 25 G
Catalog No. 502268247
Encompass_Preferred
p-Aminobenzoic acid is widely used as an intermediate in the synthesis of dyes, pharmaceuticals, and other organic compounds. It is also used as an ultraviolet (UV) absorbent in sunscreens, cosmetics and other personal care products to protect skin from harmful UV rays.In biological systems, p-aminobenzoic acid is a precursor to folic acid and plays a role in the metabolism of tyrosine, an amino acid. It is also involved in the biosynthesis of other important compounds like porphyrins, histamine, and melatonin.
Catalog No. 50-226-8247 Supplier AG Scientific Inc Supplier No. A280725GM
p-Aminobenzoic acid is widely used as an intermediate in the synthesis of dyes, pharmaceuticals, and other organic compounds. It is also used as an ultraviolet (UV) absorbent in sunscreens, cosmetics and other personal care products to protect skin from harmful UV rays.In biological systems, p-aminobenzoic acid is a precursor to folic acid and plays a role in the metabolism of tyrosine, an amino acid. It is also involved in the biosynthesis of other important compounds like porphyrins, histamine, and melatonin.