Learn More
Alkali Scientific p Aminobenzoic Acid [Vitamin H1], 100 G
SDP
Catalog No. 5000659990
Encompass_Preferred
Aminobenzoic Acid (Vitamin H1), 100g is an essential biochemical reagent used in various research applications. This compound, also known as p-aminobenzoic acid, is a precursor to folic acid and is important for the biosynthesis of purines and other essential molecules in living organisms. The 100g format is ideal for larger scale laboratory applications, including microbial culture media preparation, biochemical research, and pharmaceutical testing. It is used in the synthesis of folate derivatives and as a key ingredient in growth media for various microorganisms. Aminobenzoic acid also plays a role in enzyme activity, making it essential in a wide variety of biochemical and molecular studies.
Catalog No. 50-006-59990 Supplier Alkali Scientific Supplier No. PAM001CP
Add to Cart
Add to Cart
Aminobenzoic Acid (Vitamin H1), 100g is an essential biochemical reagent used in various research applications. This compound, also known as p-aminobenzoic acid, is a precursor to folic acid and is important for the biosynthesis of purines and other essential molecules in living organisms. The 100g format is ideal for larger scale laboratory applications, including microbial culture media preparation, biochemical research, and pharmaceutical testing. It is used in the synthesis of folate derivatives and as a key ingredient in growth media for various microorganisms. Aminobenzoic acid also plays a role in enzyme activity, making it essential in a wide variety of biochemical and molecular studies.