Learn More
Alkali Scientific p Aminobenzoic Acid [Vitamin H1], 25 G
SDP
Catalog No. 5000661307
Encompass_Preferred
Aminobenzoic Acid (Vitamin H1), 25g is a vital reagent used in biochemical and pharmaceutical research. This compound serves as a precursor to folic acid, which is essential for DNA synthesis and cellular function. Aminobenzoic acid is commonly used in microbial culture media and as a key ingredient in the preparation of growth media for microorganisms, particularly in the study of folate metabolism. It also has applications in enzyme research, metabolic pathway studies, and other biochemical applications. The 25g quantity is suitable for small-scale laboratory experiments and research studies involving the synthesis of folate derivatives and other related compounds.
Catalog No. 50-006-61307 Supplier Alkali Scientific Supplier No. PAM002CP
Aminobenzoic Acid (Vitamin H1), 25g is a vital reagent used in biochemical and pharmaceutical research. This compound serves as a precursor to folic acid, which is essential for DNA synthesis and cellular function. Aminobenzoic acid is commonly used in microbial culture media and as a key ingredient in the preparation of growth media for microorganisms, particularly in the study of folate metabolism. It also has applications in enzyme research, metabolic pathway studies, and other biochemical applications. The 25g quantity is suitable for small-scale laboratory experiments and research studies involving the synthesis of folate derivatives and other related compounds.