Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β(p-Aminophenyl)propionic Acid, MP Biomedicals

Catalog No. ICN226675 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226675 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226675 Supplier MP Biomedicals, Inc Supplier No. 0522667501

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2393-17-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key WXOHKMNWMKZMND-UHFFFAOYSA-N
Synonym 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid
PubChem CID 75451
IUPAC Name 3-(4-aminophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)N

Specifications

Quantity 10 g
Formula Weight 165.19
Chemical Name or Material β-(p-Aminophenyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.