Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Aminophenylarsonic Acid, MP Biomedicals

Catalog No. ICN203968 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203968 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203968 Supplier MP Biomedicals, Inc Supplier No. 0520396880

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-50-0
Molecular Formula C6H8AsNO3
Molecular Weight (g/mol) 217.056
InChI Key XKNKHVGWJDPIRJ-UHFFFAOYSA-N
Synonym p-arsanilic acid, arsanilic acid, 4-arsanilic acid, 4-aminophenyl arsonic acid, 4-aminobenzenearsonic acid, atoxylic acid, p-aminobenzenearsonic acid, aminophenylarsine acid, p-anilinearsonic acid, pro-gen
PubChem CID 7389
ChEBI CHEBI:49477
IUPAC Name (4-aminophenyl)arsonic acid
SMILES C1=CC(=CC=C1N)[As](=O)(O)O

Specifications

Quantity 100 g
Formula Weight 217.06
Chemical Name or Material p-Aminophenylarsonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.