Learn More
Medchemexpress LLC p-Aminophenylmercuric acetate | 6283-24-5 | 99.1% | 351.75 | 1 G
SDP
Catalog No. 5000360800
Encompass_Preferred
p-Aminophenylmercuric acetate is an organomercurial activator of matrix metalloproteinases (MMP). It participates in the activation and inhibition of MMP-8 by attacking protein sulfhydryl or inducing cysteine switching reactions. It also promotes the shedding of betacellulin precursor (pro-BTC) and influences the binding of agonists and antagonists to the opiate receptor.
  • Organomercurial activator of matrix metalloproteinases (MMP)
  • Activates MMP-2 and MMP-9 at 0.5 mM for 30 minutes
  • Decreases the apparent number of dihydromorphine binding sites and increases the sensitivity of agonist binding to the inhibitory effects of sodium in rat brain homogenate at 0-30 μM for 20 minutes
  • Promotes the shedding of betacellulin precursor (pro-BTC)
  • Influences the binding of agonists and antagonists to the opiate receptor
  • Participates in the activation and inhibition of MMP-8
Catalog No. 50-003-60800 Supplier Medchemexpress LLC Supplier No. HY1489051G
Add to Cart
Add to Cart
p-Aminophenylmercuric acetate is an organomercurial activator of matrix metalloproteinases (MMP). It participates in the activation and inhibition of MMP-8 by attacking protein sulfhydryl or inducing cysteine switching reactions. It also promotes the shedding of betacellulin precursor (pro-BTC) and influences the binding of agonists and antagonists to the opiate receptor.
  • Organomercurial activator of matrix metalloproteinases (MMP)
  • Activates MMP-2 and MMP-9 at 0.5 mM for 30 minutes
  • Decreases the apparent number of dihydromorphine binding sites and increases the sensitivity of agonist binding to the inhibitory effects of sodium in rat brain homogenate at 0-30 μM for 20 minutes
  • Promotes the shedding of betacellulin precursor (pro-BTC)
  • Influences the binding of agonists and antagonists to the opiate receptor
  • Participates in the activation and inhibition of MMP-8