Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Aminosalicylic Acid Sodium Salt Dihydrate, approx. 99%, MP Biomedicals™

Catalog No. ICN15036790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15036790 500 g
ICN15036725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15036790 Supplier MP Biomedicals, Inc Supplier No. 0215036790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6018-19-5
Molecular Formula C7H10NNaO5
Molecular Weight (g/mol) 211.149
InChI Key GMUQJDAYXZXBOT-UHFFFAOYSA-M
Synonym sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp
PubChem CID 16211148
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
SMILES C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+]

Specifications

Color White
Assay Percent Range approx. 99%
Quantity 500 g
Formula Weight 211.15
Physical Form Solid
Chemical Name or Material p-Aminosalicylic Acid Sodium Salt Dihydrate
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.