Learn More
ABclonal Technology p-AMPKa1-T183-AMPKa2-T172
SDP
Supplier:  ABclonal Technology AP144120UL
Encompass_Preferred
The protein encoded by this gene belongs to the ser thr protein kinase family It is the catalytic subunit of the 5 -prime-AMP-activated protein kinase (AMPK) AMPK is a cellular energy sensor conserved in all eukaryotic cells The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP ATP ratio AMPK regulates the activities of a number of key metabolic enzymes through phosphorylation It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways Alternatively spliced transcript variants encoding distinct isoforms have been observed
Catalog No. 50-000-14060