Learn More
Selleck Chemical LLC p-Anisaldehyde S5086-25ul
SDP
Catalog No. 503112135
Encompass_Preferred
p-Anisaldehyde (4-Methoxybenzaldehyde Anisaldehyde Anisic aldehyde P-Methoxybenzaldehyde 4-Anisaldehyde) an extract from Pimpinella anisum seeds exhibits antifungal activity against a number of yeast and mold strains in laboratory media fruit purees and fruit juices
Catalog No. 50-311-2135 Supplier Selleck Chemical LLC Supplier No. S508625UL
p-Anisaldehyde (4-Methoxybenzaldehyde Anisaldehyde Anisic aldehyde P-Methoxybenzaldehyde 4-Anisaldehyde) an extract from Pimpinella anisum seeds exhibits antifungal activity against a number of yeast and mold strains in laboratory media fruit purees and fruit juices